Cerex® PSCX 96 Well Plate, RCH
Lieferant: Tecan
687-0101RCH
TCAT687-0101RCHEA 0 EUR
TCAT687-0101RCH
Cerex® PSCX 96 Well Plate, RCH
Mikrotiterplatten Festphasenextraktionsplatten
A 25 µm polymer micro particulate with strong cationic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep.
  • Cerex
  • Ion-exchange
  • Liquid handler compatible
Ideal for basic target compounds.
Verpackung: 1 piece per box