Cerex® CLIN II SPE Column
Lieferant: Tecan
691-0353C
TCAT691-0353CEA 0 EUR
TCAT691-0353C
Cerex® CLIN II SPE Column
Chromatographiesäulen
Micro particulate with strong cation exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample preparation. Formulated for FNT and pain panel.