Microplate reader, Infinite® F50 Plus
Lieferant: Tecan
91000001
TCAT91000001EA 0 EUR
TCAT91000001
Microplate reader, Infinite® F50 Plus
Mikrotiterplatten-Photometer
The Tecan Infinite F50 Plus absorbance plate reader, together with the Magellan data analysis software, is an ideal solution for photometric measurements of ELISAs and kinetic assays in the 400 to 750 nm wavelength range.
  • Ideal for absorbance-based ELISAs, kinetic and multi-label reading
  • 405, 450, 492 and 620 nm filters included
  • Ready for qualitative and quantitative data analysis using Magellan data analysis software
  • 8-channel optics + reference channel for fast reading & optimized light intensity
  • Linear plate shaking
  • Designed to meet IVDR (EU) 2017/746
The Infinite F50 Plus is a reliable, validated and compact absorbance microplate reader for ELISA. The reader provides accurate, fast photometric measurements using innovative eight-channel optics and LED technology.
Together with Magellan™ reader control and data analysis software, this reader is ideal for a variety of ELISA applications. The compact Infinite F50 Plus fits into any lab, saving valuable bench space.
Two year warranty and tested reliability of >100,000 plates.
Bestellinformation: Accessories not included (must be ordered seperately). Additional configuration available on request.
Lieferumfang: Includes 1× reader, 1× Magellan data analysis software, 1× power cable, 1× USB stick with manuals in 11 languages.