Microplate washer, HydroFlex™ Plus
Lieferant: Tecan
91100001
TCAT91100001EA 0 EUR
TCAT91100001
Microplate washer, HydroFlex™ Plus
Mikrotiterplatten-Washer
The HydroFlex Plus microplate washer is a truly flexible and upgradeable platform for automated microplate strip washing n 96-well plate formats. It is an ideal solution for a wide range of cell- and ELISA-based applications.
  • Control of wash head positions and speed settings for gentle cell washing
  • Simultaneous aspiration and dispensing for ELISA washing
  • Up to two aspiration points per well for minimal residual volumes
  • Ready for robotic integration
  • Automated buffer switching of four liquids and plate shaking capability
  • Predefined plate library and protocols for an easy start
  • Designed to meet IVDR (EU) 2017/746 (only for ELISA wash)
The HydroFlex Plus offers both cell and ELISA washing, without the need for dedicated wash heads. It provides consistent, high- quality washing with very low residual volumes of less than 2 μl per well, and features multiple aspiration points for flat-bottom well geometries.
The overflow function offers excellent single-step washing effciency, avoiding the need for tedious repositioning of the wash head for sequential aspiration and dispense steps.
Bestellinformation: Accessories not included (must be ordered seperate). Additional configuration available on request.
Lieferumfang: Includes 1× Washer, 1× 5 L set bottle waste, 2× 2,5 L wash bottles, 1× power cable, 1× USB stick with manuals in 11 languages.