Microplate reader, Sunrise®
Lieferant: Tecan
95000001
TCAT95000001EA 0 EUR
TCAT95000001
Microplate reader, Sunrise®
Mikrotiterplatten-Photometer
This Sunrise absorbance microplate reader offers the functionality needed for numerous photometric applications.
  • Ideal for colometric ELISA, kinetic assays and agglutination readings
  • Temperature control option for enzyme kinetics
  • Advanced 12-channel optics + reference channel for fast and optimised results
  • Microplate shaking
  • Ready for robotic integration
The Sunrise absorbance reader, complemented by Tecan’s Magellan™ universal reader control and data analysis software add-on, is an ideal solution for a range of applications, such as ELISAs, enzyme kinetics, cell viability studies, veterinary assays and many more.
It supports a broad wavelength range – from 340 to 750 nm – and can measure an entire 96-well plate in less than si× seconds, ensuring it is ready to meet the needs of routine research labs. The three-year warranty demonstrate Tecan’s commitment to quality.
Bestellinformation: Accessories not included (must be ordered seperately). Additional configuration available on request.
Lieferumfang: Includes 1× reader, 1× data analysis software 1× power cable, 1× USB stick with manuals in 11 languages.