9-Acridon 98,0 HPLC, N
Lieferant: TCI
|
Achtung
|
Synonyme:
9,10-Dihydro-9-oxoacridin, 9(10H)-Acridanone, Acridone, 9(10H)-Acridone, Acridin-9(10H)-one, 9-Acridone
A0133-25G
A0133-5G
TCIAA0133-25GEA
284
EUR
TCIAA0133-25G
TCIAA0133-5G
9-Acridon 98,0 HPLC, N
9-Acridon
Formel:
C₁₃H₉NO Molecular Weight: 195.22 g/mol Schmelzpunkt: 354°C |
MDL:
MFCD00005019 CAS-Nummer: 578-95-0 EINECS: 209-434-7 |
|
Spezifikation Testergebnisse
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...