Acriflavin ≥70% (durch HPLC) (mixture of 3,6-Diamino-10-methylacridinium Chloride and 3,6-Diaminoacridine)
Lieferant: TCI
|
Achtung
|
Synonyme:
3,6-Diamino-10-methylacridinium chloride, 3,6-Diamino-10-methylacridinium chloride-3,6-acridinediamine mix, Acriflavinium chloride 3,6-Acridinediamine mix, 3,6-Diamino-10-methylacridinium chloride mixt. with 3,6-diaminoacridine (proflavine), Euflavine, Trypaflavine Neutral, Euflavine, Acriflavine base
A0134-25G
TCIAA0134-25GEA
122
EUR
TCIAA0134-25G
Acriflavin ≥70% (durch HPLC) (mixture of 3,6-Diamino-10-methylacridinium Chloride and 3,6-Diaminoacridine)
Acriflavin
Formel:
C₂₇H₂₅ClN₆ Molecular Weight: 468,9806 g/mol |
MDL:
MFCD00064307 CAS-Nummer: 8048-52-0 Merck Index: 14,00123 |
Spezifikation Testergebnisse
Appearance | Orange to Amber to Dark red powder to crystal |
Purity (HPLC) | min. 70.0 % (as Acriflavine) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...