6,9-Diamino-2-ethoxyacridine lactate monohydrate ≥98,0% (durch HPLC, titrimetrische Analyse)
Lieferant: TCI
|
Achtung
|
Synonyme:
Ethodin, 7-Ethoxyacridine-3,9-diamine 2-hydroxypropanoate hydrate, Ethacridine lactate salt, Acrinol Monohydrate
A0136-25G
TCIAA0136-25GEA
104
EUR
TCIAA0136-25G
6,9-Diamino-2-ethoxyacridine lactate monohydrate ≥98,0% (durch HPLC, titrimetrische Analyse)
6,9-Diamino-2-ethoxyacridine lactate monohydrate
Formel:
C₁₈H₂₃N₃O₅ |
MDL:
MFCD00149646 CAS-Nummer: 6402-23-9 Merck Index: 14,03716 |
Spezifikation Testergebnisse
Appearance | Light yellow to Yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % (calcd.on anh.substance) |
Solubility in hot Water | almost transparency |
Water | 4.0 to 6.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...