Acrylamide polymer, M.W. 400000-800000 10% water (containing small amounts of formalin as fungicide)
Lieferant: TCI
Synonyme:
2-Propenamide Homopolymer, poly(1-carbamoylethylene), Polyacrylamide
A0140-500G
TCIAA0140-500GEA
213
EUR
TCIAA0140-500G
Acrylamide polymer, M.W. 400000-800000 10% water (containing small amounts of formalin as fungicide)
Acrylamide polymer
Formel:
(CH₂:CHCONH₂)n Lagertemperatur: Kühlschrank |
MDL:
MFCD00084392 CAS-Nummer: 9003-05-8 |
Spezifikation Testergebnisse
Appearance | Colorless to Almost colorless clear liquid |
Molecular weight | 400,000 to 800,000 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...