Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz) Hydrat ≥98,0% (durch HPLC, titrimetrische Analyse)

Lieferant: TCI

Synonyme: ATP disodium salt hydrate, 5′-ATP-Na2 ATP disodium salt hydrate, Adenosine 5'-Triphosphate Disodium Salt Hydrate, Adenosine-5'-triphosphate disodium salt hydrate, 5'-ATP 2Na Hydrate, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate disodium salt hydrate, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt xhydrate, Adenosine 5'-triphosphoric acid disodium hydrate

A0157-25G A0157-1G A0157-5G
TCIAA0157-25GEA 359 EUR
TCIAA0157-25G TCIAA0157-1G TCIAA0157-5G
Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz) Hydrat ≥98,0% (durch HPLC, titrimetrische Analyse)
Adenosin-5'-triphosphat Dinatriumsalz (ATP Dinatriumsalz) Hydrat
Formel: C₁₀H₁₄N₅Na₂O₁₃P₃·xH₂O
Lagertemperatur: Kühlschrank
MDL: MFCD00080339
CAS-Nummer: 34369-07-8
EINECS: 213-579-1
Merck Index: 14,00155
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be5ac448451f84b10996626c6c8626bd970bd2ea&vid=be5ac448451f84b10996626c6c8626bd970bd2ea&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
abs-ratio (a/b) 0.78 to 0.82 (E250/E260)
abs-ratio (2a/2b) 0.13 to 0.17 (E280/E260)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR