Adenosine-3',5'-cyclic monophosphate sodium salt ≥98,0% (durch HPLC)

Lieferant: TCI

Synonyme: 3′,5′-Cyclic AMP sodium salt, 3',5'-Cyclic AMP Sodium Salt, Adenosine 3',5'-Cyclic Monophosphate Sodium Salt Hydrate, 3',5'-cAMP Na, (4aR,6R,7R,7aS)-6-(6-Amino-9H-purin-9-yl)-2,7-dihydroxytetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinine 2-oxide, sodium salt, cAMP-Na, 3',5'-Cyclic AMP Sodium Salt Hydrate, 3',5'-cAMP Na Hydrate

A2112-100MG
TCIAA2112-100MGEA 121 EUR
TCIAA2112-100MG
Adenosine-3',5'-cyclic monophosphate sodium salt ≥98,0% (durch HPLC)
Adenosine-3',5'-cyclic monophosphate sodium salt
Formel: C₁₀H₁₁N₅NaO₆P
Molecular Weight: 351,19 g/mol
MDL: MFCD00069736
CAS-Nummer: 37839-81-9

Bestellen

Spezifikation Testergebnisse

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Water max. 10.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR