2-Aminoadenosin ≥98,0% (durch HPLC, titrimetrische Analyse)
Lieferant: TCI
|
Achtung
|
Synonyme:
2,6-Diamino-9-(beta-D-ribofuranosyl)purine, (2R,3R,4S,5R)-2-(2,6-Diamino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, 2,6-Diaminopurine Riboside
A2135-25G
A2135-5G
TCIAA2135-25GEA
403
EUR
TCIAA2135-25G
TCIAA2135-5G
2-Aminoadenosin ≥98,0% (durch HPLC, titrimetrische Analyse)
2-Aminoadenosine
Formel:
C₁₀H₁₄N₆O₄ Molecular Weight: 282.26 g/mol Schmelzpunkt: 252°C |
MDL:
MFCD00053556 CAS-Nummer: 2096-10-8 |
Spezifikation Testergebnisse
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | -36.0 to -44.0 deg (C=1, 0.1mol/L HCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...