(4-Amino-2-methyl-5-pyrimidinyl)methanol ≥98,0% (durch GC, titrimetrische Analyse)
|
Achtung
|
Synonyme:
Toxopyrimidine, 4-Amino-2-methyl-5-pyrimidinemethanol, 4-Amino-5-hydroxymethyl-2-methylpyrimidine
A2730-200MG
A2730-1G
TCIAA2730-200MGEA
124
EUR
TCIAA2730-200MG
TCIAA2730-1G
(4-Amino-2-methyl-5-pyrimidinyl)methanol ≥98,0% (durch GC, titrimetrische Analyse)
(4-Amino-2-methyl-5-pyrimidinyl)methanol
Formel:
C₆H₉N₃O Molecular Weight: 139.1552 g/mol |
MDL:
MFCD00849239 CAS-Nummer: 73-67-6 Merck Index: 14,09561 |
Spezifikation Testergebnisse
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 195.0 to 199.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...