Sodium-3-bromopropanesulphonate ≥98,0% (durch titrimetrische Analyse)
|
Achtung
|
Synonyme:
3-Bromopropanesulphonic acid Sodium Salt
B1707-25G
B1707-100G
TCIAB1707-25GEA
126
EUR
TCIAB1707-25G
TCIAB1707-100G
Sodium-3-bromopropanesulphonate ≥98,0% (durch titrimetrische Analyse)
Sodium-3-bromopropanesulphonate
Formel:
C₃H₇BrO₃S Molecular Weight: 225,03 g/mol |
MDL:
MFCD00024999 CAS-Nummer: 55788-44-8 |
Spezifikation Testergebnisse
Appearance | White to Light yellow powder to crystal |
Purity (Ion exchange titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...