2',3'-Didesoxyadenosin ≥98,0% (durch HPLC, titrimetrische Analyse)

Lieferant: TCI
Achtung

Synonyme: ((2S,5R)-5-(6-Amino-9H-purin-9-yl)tetrahydrofuran-2-yl)methanol

D3065-100MG
TCIAD3065-100MGEA 537 EUR
TCIAD3065-100MG
2',3'-Didesoxyadenosin ≥98,0% (durch HPLC, titrimetrische Analyse)
2',3'-Didesoxyadenosin
Formel: C₁₀H₁₃N₅O₂
Molecular Weight: 235,24 g/mol
MDL: MFCD00010534
CAS-Nummer: 4097-22-7
EINECS: 223-853-2
Merck Index: 14,03101
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5320abaef86175784588c3bc5f9fb8fedfb35d10&vid=5320abaef86175784588c3bc5f9fb8fedfb35d10&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 186.0 to 190.0 deg-C
Specific rotation [a]20/D -27.0 to -34.0 deg (C=1, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR