2'-Desoxyadenosin, wasserfrei ≥98,0% (durch HPLC, titrimetrische Analyse)
Lieferant: TCI
Synonyme:
(2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol
D4137-5G
TCIAD4137-5GEA
193
EUR
TCIAD4137-5G
2'-Desoxyadenosin, wasserfrei ≥98,0% (durch HPLC, titrimetrische Analyse)
2'-Desoxyadenosin
Formel:
C₁₀H₁₃N₅O₃ Molecular Weight: 251,24 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00005754 CAS-Nummer: 958-09-8 EINECS: 213-488-7 |
Spezifikation Testergebnisse
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 188.0 to 192.0 deg-C |
Specific rotation [a]20/D | -25.0 to -28.0 deg (C=0.5, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...