Dibutyryl-cAMP sodium salt ≥97,0% (durch HPLC)
Lieferant: TCI
Synonyme:
Bucladesine sodium salt, DC-2797, Dibutyl cyclic AMP sodium salt, Dibutyryl-cAMP, N6,2'-O-Dibutyryladenosine 3',5'-Cyclic monophosphate sodium salt, DbcAMP, Sodium (4aR,6R,7R,7aR)-6-(6-butyramido-9H-purin-9-yl)-7-(butyryloxy)tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-2-olate 2-oxide, Bucladesine
D4228-25MG
TCIAD4228-25MGEA
161
EUR
TCIAD4228-25MG
Dibutyryl-cAMP sodium salt ≥97,0% (durch HPLC)
Dibutyryl-cAMP sodium salt
Formel:
C₁₈H₂₃N₅O₈P·Na Molecular Weight: 491,37 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00005843 CAS-Nummer: 16980-89-5 EINECS: 241-059-4 Merck Index: 14,01463 |
Spezifikation Testergebnisse
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Elemental analysis (Nitrogen) | 13.40 to 14.60 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...