3-(2-Ethylhexyloxy)-1,2-propanediol ≥98,0% (durch GC)
|
Gefahr
|
Synonyme:
Glycerol alpha-(2-Ethylhexyl) Ether, 3-((2-Ethylhexyl)oxy)propane-1,2-diol
E1044-25ML
TCIAE1044-25MLEA
117
EUR
TCIAE1044-25ML
3-(2-Ethylhexyloxy)-1,2-propanediol ≥98,0% (durch GC)
3-(2-Ethylhexyloxy)-1,2-propanediol
Formel:
C₁₁H₂₄O₃ Molecular Weight: 204.31 g/mol Flammpunkt: 152°C |
MDL:
MFCD03790878 CAS-Nummer: 70445-33-9 EINECS: 408-080-2 |
Spezifikation Testergebnisse
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...