3-Fluor-N-methylbenzylamin
Lieferant: TCI
Synonyme:
(3-Fluorobenzyl)methylamine, 1-(3-Fluorophenyl)-N-methylmethanamine
F0805-1G
TCIAF0805-1GEA
52.8
EUR
TCIAF0805-1G
3-Fluor-N-methylbenzylamin
3-Fluor-N-methylbenzylamin
Formel:
C₈H₁₀FN Molecular Weight: 139,17 g/mol |
MDL:
MFCD04623547 CAS-Nummer: 90389-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...