Calcium-3-hydroxy-3-methylbutyrate hydrate ≥97,0% (durch titrimetrische Analyse)

Synonyme: 3-Hydroxy-3-methylbutyric Acid Calcium Salt

H0917-25G
TCIAH0917-25GEA 67.4 EUR
TCIAH0917-25G
Calcium-3-hydroxy-3-methylbutyrate hydrate ≥97,0% (durch titrimetrische Analyse)
Calcium-3-hydroxy-3-methylbutyrate hydrate
Formel: C₁₀H₁₈CaO₆·H₂O
Molecular Weight: 274,33 g/mol (anhy)
MDL: MFCD01321171
CAS-Nummer: 135236-72-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b4357cd1d37273562eb101105b947cf5fb86e61&vid=04cc17511f24056ea1e9d434f1ac25963abafb41&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 97.0 % (calcd.on anh.substance)
Solubility in Water very faint turbidity
Water max. 10.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR