(4-Hydroxyphenyl)phosphonic acid ≥97,0% (durch HPLC, titrimetrische Analyse)

Achtung

H1559-200MG H1559-1G
TCIAH1559-200MGEA 170 EUR
TCIAH1559-200MG TCIAH1559-1G
(4-Hydroxyphenyl)phosphonic acid ≥97,0% (durch HPLC, titrimetrische Analyse)
(4-Hydroxyphenyl)phosphonic acid
Formel: C₆H₇O₄P
Molecular Weight: 174.09 g/mol
MDL: MFCD01112120
CAS-Nummer: 33795-18-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1e6c977b78fb66a1c4f5305cb483fe0ee897ee0&vid=b1e6c977b78fb66a1c4f5305cb483fe0ee897ee0&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR