1-Methylaminomethylnaphthalin 98,0 Titrimetric

Achtung

Synonyme: 1-(Methylaminomethyl)naphthalene

M0983.1 M0983-5G
TCIAM0983.1EA 66.5 EUR
TCIAM0983.1 TCIAM0983-5G
1-Methylaminomethylnaphthalin 98,0 Titrimetric
1-Methylaminomethylnaphthalin
Formel: C₁₂H₁₃N
Molecular Weight: 171.24 g/mol
Siedepunkt: 120°C
MDL: MFCD00144934
CAS-Nummer: 14489-75-9
EINECS: 238-497-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=daa05759d85508f9a4e9aa0822e9f5b89b2de875&vid=daa05759d85508f9a4e9aa0822e9f5b89b2de875&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to Orange to Green clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR