Malonaldehyde dianilide hydrochloride ≥98,0% (durch HPLC, titrimetrische Analyse)
|
Achtung
|
Synonyme:
Malonaldehyde dianilide xHydrochloride, N-(3-(Phenylamino)allylidene)aniline hydrochloride, Malonaldehyde bis(phenylimine) hydrochloride
M1288-25G
TCIAM1288-25GEA
81.9
EUR
TCIAM1288-25G
Malonaldehyde dianilide hydrochloride ≥98,0% (durch HPLC, titrimetrische Analyse)
Malonaldehyde dianilide hydrochloride
Formel:
C₁₅H₁₄N₂ Molecular Weight: 258.75 g/mol |
MDL:
MFCD00054634 CAS-Nummer: 28140-60-5 EINECS: 248-862-9 |
Spezifikation Testergebnisse
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...