7-Methoxycoumarin-3-carboxylic acid ≥98,0% (durch GC, titrimetrische Analyse)
|
Achtung
|
Synonyme:
7-Methoxy-2-oxo-2H-chromene-3-carboxylic acid
M2233-1G
M2233-100MG
TCIAM2233-1GEA
314
EUR
TCIAM2233-1G
TCIAM2233-100MG
7-Methoxycoumarin-3-carboxylic acid ≥98,0% (durch GC, titrimetrische Analyse)
7-Methoxycoumarin-3-carboxylic acid
Formel:
C₁₁H₈O₅ Molecular Weight: 220.18 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00452770 CAS-Nummer: 20300-59-8 |
Spezifikation Testergebnisse
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 194.0 to 198.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...