2-(9-Oxoxanthen-2-yl)propionic acid 1,5-Diazabicyclo[4.3.0]non-5-ene salt ≥98,0% (durch HPLC)

Achtung

O0447-1G
TCIAO0447-1GEA 492 EUR
TCIAO0447-1G
2-(9-Oxoxanthen-2-yl)propionic acid 1,5-Diazabicyclo[4.3.0]non-5-ene salt ≥98,0% (durch HPLC)
2-(9-Oxoxanthen-2-yl)propionic acid 1,5-Diazabicyclo[4.3.0]non-5-ene salt
Formel: C₂₃H₂₄N₂O₄
Molecular Weight: 392,46 g/mol
CAS-Nummer: 1346753-04-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c22d45c581fb5c17b1ff80c1e4105b890b548c75&vid=c22d45c581fb5c17b1ff80c1e4105b890b548c75&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 % (total of Ion-Pair)
Elemental analysis (Nitrogen) 7.00 to 7.20 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR