1-({5-[(3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanoyl}oxy)-2,5-pyrrolidindion ≥97,0% (durch HPLC, Gesamtstickstoff)

Synonyme: Biotin-NHS, 2,5-Dioxopyrrolidin-1-yl 5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoate, (+)-Biotin N-hydroxysuccinimide ester, D-Biotinyl-OSu, NHS-Biotin, Biotin N-hydroxysuccinimide ester, N-(D-Biotinyloxy)succinimide

S0491-100MG S0491-1G
TCIAS0491-100MGEA 34.6 EUR
TCIAS0491-100MG TCIAS0491-1G
1-({5-[(3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanoyl}oxy)-2,5-pyrrolidindion ≥97,0% (durch HPLC, Gesamtstickstoff)
D-Biotin-N-succinimidyl ester
Formel: C₁₄H₁₉N₃O₅S
Molecular Weight: 341,38 g/mol
Schmelzpunkt: 140°C
Lagertemperatur: Gefrierschrank
MDL: MFCD00078531
CAS-Nummer: 35013-72-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ccde0dab244f5d9817f201bd09425300361cc60&vid=3ccde0dab244f5d9817f201bd09425300361cc60&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Protein labeling test (qualitative methoed) to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR