Cole-Parmer® GT-200 Gelation Timers
Lieferant: Cole-Parmer
98930-41 98930-43 98930-49 98930-47 98930-48 98930-46 98930-44
TECN98930-41EA 3050 EUR
TECN98930-41 TECN98930-43 TECN98930-49 TECN98930-47 TECN98930-48 TECN98930-46 TECN98930-44
Cole-Parmer® GT-200 Gelation Timers
Measure the gelation time of adhesives and resins for quality control.
  • Automatically stops when gelation occurs
  • Audible and visual alarms alert user at the gelation point
  • Simple-to-use push buttons with large LED display
  • Stainless steel plungers remove for easy cleaning
The gelation time is the transition from liquid to solid during polymerisation. Test requires approximately 100 ml of sample in a container that is >1,5" (38 mm) in diameter. Once the measurement is started, the plunger moves up and down. After gelation, the plunger is halted, stopping the timer and setting off the audible and visual alarms. The bright LED digital display shows the time to the nearest minute or tenth of a minute.
Bestellinformation: Stand and clamp are sold separately.
Lieferumfang: Stainless steel paddle and power cord with stripped ends.