Cleanroom Pedestal Waste Bag Holders
Lieferant: TEKNOMEK
HWBH-OFNLCR
TEKMHWBH-OFNLCREA 0 EUR
TEKMHWBH-OFNLCR
Cleanroom Pedestal Waste Bag Holders
Behälter Abfallbehälter
Our cleanroom pedestal waste bag holders are manufactured from 304L-grade stainless steel. A plastic part free design and food grade edge trims ensure our products are easy to clean, supporting rigorous hygiene standards. A tough stainless steel clamping band securely holds the bag in place, reducing the risk of tearing or spilling waste.
  • Open or enclosed version available
  • Non-shedding clean room grade
  • Strong and sturdy design
  • Supplied ready to use
  • Foot operated lids available for easy operation
  • Etching option available
  • Ring H×Ø: 75×350 mm
Available without a lid or with foot operated lids, to aid with hands-free use, are available.
Open or enclosed versions that offer even more use ability are available whilst ensuring hygiene standards are maintained.