Bottle Holders
Lieferant: TEKNOMEK
HWBH-0500-1 HWDI-BH3B
TEKMHWBH-0500-1EA 0 EUR
TEKMHWBH-0500-1 TEKMHWDI-BH3B
Bottle Holders
Transportbehälter Flaschentragebehälter
Our bottle holders come in a range of sizes.
  • Brushed dull polish finish
  • Easy clean design
  • 80 mm diameter hole
Our bottle holders are the ideal solution to your storage needs, coming in a range of sizes (1, 2 or 3 bottle capacities) to suit your requirements.
The bottle holders are manufactured in fully welded 304 grade stainless steel with a brushed dull polish finish.
The stainless steel bottle holder is suitable for housing 500 ml bottles with 80 mm diameter rings, ensuring they can hold a wide variety of bottles.