Storage cupboards with internal cabinet
Lieferant: TEKNOMEK
SHCBIC12X60X17
TEKMSHCBIC12X60X17EA 0 EUR
TEKMSHCBIC12X60X17
Storage cupboards with internal cabinet
Möbel Lagerschränke
Our storage cupboard comes complete with an internal lockable cabinet.
  • Brushed dull polish finish
  • Internal adjustable shelf
  • Stainless steel adjustable feet with non-marking rubber base
  • Hinged lockable doors
  • Internal lockable drug storage safe
Our storage cupboard is ideal for storing pharmaceuticals safely. An internal lockable cabinet and hinged lockable external doors ensure security, while an adjustable shelf provides practical storage.
Bestellinformation: Available in two different depths and widths.