Stainless Steel Writing Desk with Undershelf
Lieferant: TEKNOMEK
TLWD-1S60X40
TEKMTLWD-1S60X40EA 0 EUR
TEKMTLWD-1S60X40
Stainless Steel Writing Desk with Undershelf
Möbel Tische und Werkbänke
This stainless steel writing desks are the ideal workspace for any industry which count hygiene and longevity as priorities. They come with lockable hinged lids and a choice of stainless steel adjustable feet or stainless steel castors for the mobile version.
  • 304L-grade stainless steel
  • Brushed dull polish finish
  • Inclined top with lockable compartment
  • Available with stainless steel feet or stainless steel castors
This robust 304L-grade stainless steel writing desks come in both static and mobile options and with a brushed dull polish finish. The desk top is inclined with a lip for secure storage of papers and is complete with a removable rubber edge trim to protect the user without compromising the clean down process.
It's intelligent and hygienic design makes our writing desks easy to clean, with flat edges underneath to avoid dirt collection and side-positioned hinges on the sloping lid to make the unit splash resistant.
The sturdy undershelf is ideal for additional storage.