Natriumazid-Lösung 10% (w/v), steril

Lieferant: TEKNOVA
Gefahr

Synonyme: Natriumtriazid

S0209
TEKOS0209EA 404 EUR
TEKOS0209
Natriumazid-Lösung 10% (w/v), steril
Natriumazid
Formel: NaN₃
Molecular Weight: 65,01 g/mol
MDL: MFCD00003536
CAS-Nummer: 26628-22-8
UN: 1687
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR