TERUFUSION™ Solution Administration Set
Lieferant: TERUMO
TIPU300WY
TERUTIPU300WYEA 1190 EUR
TERUTIPU300WY
TERUFUSION™ Solution Administration Set
Infusionen
TERUFUSION™ solution administration set (for infusion pump) has unique features meeting the various needs of hospitals.
  • Simple
  • Efficient use
  • PVC, DEHP-free
  • AFF clip, Y-site×1, round lock connector
TERUFUSION™ solution administration set for infusion pump is specially designed for use with the TERUFUSION infusion pump for optimal performance and safety.