Protective Cap
Lieferant: Testo
0554.9913
TEST0554.9913EA 66.6 EUR
TEST0554.9913
Protective Cap
Verschlüsse Verschlüsse für Reagenz- und Probenröhrchen
The protective cap made of PTFE prevents damage to the sensor system caused by humidity or moisture in processes with high humidity. Optimum protection is provided by the protective cap in combination with our condensation protector.
  • Protective cap made of PTFE with condensate drip hole 1,5 mm
  • Pore size 100 µm
  • For applications in dust-free processes
  • For flow velocities up to 10 m/s