Alpha® Micro Cleanroom Swab, Polyester Knit, Sterile
Lieferant: Texwipe
STX758B
TEXWSTX758BEA 1450 EUR
TEXWSTX758B
Alpha® Micro Cleanroom Swab, Polyester Knit, Sterile
Tupfer Reinigungstupfer
Texwipe’s Alpha® STX758B sterile micro cleanroom swab is constructed from 100% polyester knit material. Complete thermal bond construction eliminates adhesive contamination.
  • Double layer of polyester knit fabric for enhanced absorbency
  • Engineered to resist snagging and abrading, preventing the release of particles and fibers
  • Excellent chemical compatibility with a variety of solutions
  • Autoclave safe in dry heat and steam
  • Each swab is individually packaged in a peel-apart sleeve
  • Tested for endotoxins, ions, and NVR
Cleanroom manufactured, providing low levels of Non-Volatile Residues (NVRs) and ions, and made to exacting and consistent tolerances using high-precision automated processes.
Lot coded for traceability and quality control. Packaged in a silicone-free and amide-free bag.
Gamma-irradiated to a Sterility Assurance Level of 10-6 and sterile validated according to AAMI Guidelines.
Cleanroom laundered, providing ultra-low levels of particles, NVRs, and ions and trademarked light-green colored handles with 'TEXWIPE' name embossed on handle.
Triple-bagged bags are packaged in a case liner forming the fourth layer of bag protection.