Cleanroom wipes, sterile, TechniCloth®
Lieferant: Texwipe
TX3410
TEXWTX3410EA 1870 EUR
TEXWTX3410
Cleanroom wipes, sterile, TechniCloth®
Wischtücher Reinraum-Wischtücher Sterile Reinraumwischtücher
This non woven 55% cellulose/45% polyester blend wipe is an excellent general-purpose sterile wipe in areas where economy and cleanliness are of critical importance.
  • Chemically resistant to withstand exposure to common cleaning and disinfecting solutions (e.g. bleaches, phenols and quaternary)
  • Low solvent extractable levels, binder-free
  • High sorbency and strength
  • Packaging permits alcohol wipedown of exterior bag before introduction of the inner bag into sterile suites
  • Quarter-folded wipes allowing a most efficient way of cleaning
  • Small footprint for easy storage
  • Gamma irradiated to a Sterility Assurance Level of 10⁻⁶
Designed for cleaning and polishing equipment and environmental surfaces during and following production flow. Also suitable for wipedowns in sterile gowning rooms and for spill control.
Colour: White.
Verpackung: 100 quarter-folded wipes per bag. 1 stack of 20 wipers per inner bag, 5 inner bags of 20 wipers per bag, 15 bags per case.
Bitte beachten Data accurate at time of publishing, latest test results and test methods can be found on supplier's website.