Cleanroom bond paper, TexWrite® MP 10
Lieferant: Texwipe
TX5800
TEXWTX5800EA 1230 EUR
TEXWTX5800
Cleanroom bond paper, TexWrite® MP 10
Papier
TexWrite® MP 10 is a 100% synthetic substrate provides extremely low levels of particle generation along with superior usability. It is designed to use with ultraclean manufacturing environments (paperless) that cannot tolerate cellulose contamination.
  • Excellent ink capture with no risk of smearing
  • Low ESD potential for reduced risk of electrostatic damage
  • Compatible with inkjet, laser and toner-based printers and photocopiers
  • Ballpoint pens glide easily over surface, without skips or jumps
  • Autoclavable and recyclable as a other types of plastic
  • Distinctive blue coating on one side
Verpackung: 100 sheets per ream and 5 ream per box.