Base model without HEPA, without Accessories, for dry recovery only Air flow: 187 m3/h Vacuum pressure: 4572 mmH2O Recovery Capacity:23L - Dry recovery 28L Wet Dimensions: H81cmW48cmL61cm Noise level: 70 dB (A) 1 * 1 ST
Lieferant: TIGER-VAC
Base model without HEPA, without Accessories, for dry recovery only Air flow: 187 m3/h Vacuum pressure: 4572 mmH2O Recovery Capacity:23L - Dry recovery 28L Wet Dimensions: H81cmW48cmL61cm Noise level: 70 dB (A) 1 * 1 ST