Vacuum cleaner, 2D-9 NUC HEPA, Tiger-Vac®
Lieferant: TIGER-VAC
110626B
TIGV110626BEA 4870 EUR
TIGV110626B
Vacuum cleaner, 2D-9 NUC HEPA, Tiger-Vac®
Staubsauger
This unit contains a true nuclear grade HEPA Filter (H14) constructed and tested to methods consistant with ASME AG-1. An external ULPA filter (U15) is available.
  • Disposable containment drum assemblies
  • Electrically operated
  • Removable and reusable power head
Construction: Polycarbonate recovery tank.
Bitte beachten Note: For dry recovery only.