VACUUM MODEL MV-1CR S/S w/ULPA 240V FROM TIGER-VAC Technical Features: Volt: 240V, Hertz: 50Hz, Watt: 1000, Ampere: 4.2A. Dry recovery. Air Flow: 187 m3/h. Vacuum at sealed orifice: 1753 mmH2O. Recovery capacity: Dry: with Paper filter bag: 2 L. Dime 1 * 1 ST
Lieferant: TIGER-VAC
VACUUM MODEL MV-1CR S/S w/ULPA 240V FROM TIGER-VAC Technical Features: Volt: 240V, Hertz: 50Hz, Watt: 1000, Ampere: 4.2A. Dry recovery. Air Flow: 187 m3/h. Vacuum at sealed orifice: 1753 mmH2O. Recovery capacity: Dry: with Paper filter bag: 2 L. Dime 1 * 1 ST