Volts: 400V 3phase Hertz: 50Hz Ampere: 2. 7 A Watt: 1300 W. Airflow: 204 m3/h Vacuum pressure: 3300 mmH20 mmH20 Dry Recovery without paper bag: 19L Paper Bag Capacity: 15 L Dimensions: H94cmW51cmL36cm Weight: 58 kg Noise Level: 65.00 dB(A) 1 * 1 ST
Lieferant: TIGER-VAC
Volts: 400V 3phase Hertz: 50Hz Ampere: 2. 7 A Watt: 1300 W. Airflow: 204 m3/h Vacuum pressure: 3300 mmH20 mmH20 Dry Recovery without paper bag: 19L Paper Bag Capacity: 15 L Dimensions: H94cmW51cmL36cm Weight: 58 kg Noise Level: 65.00 dB(A) 1 * 1 ST