Resazurin Natriumsalz

Lieferant: Lovibond Tintometer

501800
TINT501800EA 123 EUR
TINT501800
Resazurin Natriumsalz
Resazurin Natriumsalz
Formel: C₁₂H₆NNaO₄
MDL: MFCD00005036
CAS-Nummer: 62758-13-8
EINECS: 263-718-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR