Magnetic stirrers, MIXdrive 1 XL
Lieferant: 2MAG
90600 90600_CH 90600_UK 40601
TMAG90600EA 1279 EUR
TMAG90600 TMAG90600_CH TMAG90600_UK TMAG40601
Magnetic stirrers, MIXdrive 1 XL
Rührer Magnetrührer
Ultra-flat magnetic stirrers with stainless steel housing. Operating temperatures up to 100 °C.
  • Easy to clean and firm
  • Suitable for use with highly viscous media
  • Very strong magnetic coupling by SmCo magnets, for magnetic power transmission over extended distances, for thick vessel bottoms
Equipped as standard with mixWATCH ensuring a very high process safety, e.g. when using closed non-glass vessels or for unattended overnight operation. In case of the magnetic coupling’s separation between the magnetic stirrer and the stirring bar during theMore Product Information