4-Brompyridiniumchlorid

Lieferant: TORONTO RESEARCH CHEMICALS
Achtung

Synonyme: 4-Brompyridin Hydrochlorid, 4-Brompyridin HCl

B686735-100G
TOROB686735-100GEA 1420 EUR
TOROB686735-100G
4-Brompyridiniumchlorid
4-Bromopyridinium chloride
Formel: C₅H₅BrClN
Molecular Weight: 194,46 g/mol
Schmelzpunkt: 235…238 °C
MDL: MFCD00012828
CAS-Nummer: 19524-06-2
EINECS: 243-128-4
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR