Rosuvastatin-d6 (sodium salt)

Lieferant: TORONTO RESEARCH CHEMICALS

R700502-1MG
TOROR700502-1MGEA 0 EUR
TOROR700502-1MG
Rosuvastatin-d6 (sodium salt)
Rosuvastatin-d6 (sodium salt)

Labelled rosuvastatin, a selective, competitive HMG-CoA reductase inhibitor.


  • Hygroscopic
  • Store under inert atmosphere
  • Slightly soluble in DMSO, methanol


Synonyms: 7-[4-(4-Fluorophenyl)-6-(1-methylethyl-d6)-2-[methyl(methylsulphonyl)amino]-5-pyrimidinyl]-3,5-dihydroxy-6-heptenoic Acid sodium salt.

Formel: C₂₂H₂₁D₆FN₃O₆S·Na
Schmelzpunkt: >193 °C (Zersetzung)
Lagertemperatur: Gefrierschrank
CAS-Nummer: 2070009-41-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR