Size exclusion chromatography sorbents, Toyopearl®
Lieferant: TOSOH Bioscience
0007448
0007451
0007453
0007455
0007457
0007459
0007465
0007466
0007467
0007469
0014682
0014686
0014687
0014688
0014691
0019810
0019815
0019807
0019808
0019811
0019812
0019816
0019813
0019819
0019820
0019821
0019814
0014683
0007449
0014681
0014684
0014685
0014689
0014690
0021481
TOYO0007448EA
789
EUR
TOYO0007448
TOYO0007451
TOYO0007453
TOYO0007455
TOYO0007457
TOYO0007459
TOYO0007465
TOYO0007466
TOYO0007467
TOYO0007469
TOYO0014682
TOYO0014686
TOYO0014687
TOYO0014688
TOYO0014691
TOYO0019810
TOYO0019815
TOYO0019807
TOYO0019808
TOYO0019811
TOYO0019812
TOYO0019816
TOYO0019813
TOYO0019819
TOYO0019820
TOYO0019821
TOYO0019814
TOYO0014683
TOYO0007449
TOYO0014681
TOYO0014684
TOYO0014685
TOYO0014689
TOYO0014690
TOYO0021481
Size exclusion chromatography sorbents, Toyopearl®
Chromatographie-Sorbentien
HPLC-Sorbentien
Toyopearl resins can be used in organic solvents or mixtures of organic solvents and water. The compatibility of DMF with Toyopearl also permits the separation of hydrophobic substances such as polystyrenes.
- Removal of surfactants such as Triton® X-100 from biological solutions by an adsorption mechanism
- Use in hydrophobic interaction chromatography (HIC) for the separation of very hydrophobic molecules
- Suitable for HIC separations as a guard column for hydrophobic impurities
- Used as a stationary phase for either normal or reversed phase separations depending on solvent system selected
The Toyopearl HW-type resins are commonly used in size exclusion chromatography and desalting applications.