Disposable weighing boats, PP
Lieferant: TRADEWINDS DIRECT
ASWF4XL
TRADASWF4XLEA 0 EUR
TRADASWF4XL
Disposable weighing boats, PP
Schiffchen Wägeschiffchen
PP, disposable.
  • Extra large size with smooth surfaces
  • Neck (Ø= 19 mm) fits into 1000, 2000 ml volumetric flasks
Ideal for weighing solid and liquid products.