3-Aminobenzamid

Lieferant: TREVIGEN

Synonyme: 3-Aminobenzoesäureamid

4667-50-03
TREV4667-50-03EA 0 EUR
TREV4667-50-03
3-Aminobenzamid
3-Aminobenzamid
Formel: C₇H₈N₂O
Molecular Weight: 136,15 g/mol
Siedepunkt: 341 °C (1013 hPa)
Schmelzpunkt: 111…115 °C
MDL: MFCD00007989
CAS-Nummer: 3544-24-9
EINECS: 222-586-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR