Propidiumiodid

Lieferant: TREVIGEN
Achtung

Synonyme: 3,8-Diamino-5-(3-(diethylmethylammonio)propyl)-6-phenylphenanthridiniumdiiodid

4830-01-03 4830-250-03
TREV4830-01-03EA 0 EUR
TREV4830-01-03 TREV4830-250-03
Propidiumiodid
Propidiumiodid
Formel: C₂₇H₃₄I₂N₄
MDL: MFCD00011921
CAS-Nummer: 25535-16-4
EINECS: 247-081-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR