Manual detergent, alkaline, Tanzanite
Lieferant: TRISTEL SOLUTIONS
CRY304
TRSTCRY304EA 195 EUR
TRSTCRY304
Manual detergent, alkaline, Tanzanite
Reinigungsmittel Waschmittel und Oberflächenreiniger
A medium foam, heavy duty, detergent for removal of organic soils, including fats, oils and silicone.
  • Heavy-duty alkaline detergent for organic soils
  • Dilution for use: between 1 to 5% v/v
Areas of use include walls, floors, product contact areas and equipment such as vessels, filling lines and utensils. Tanzanite is not soft metal safe, but can be used on a variety of surfaces including stainless steel.
All detergents are supported by comprehensive technical files which provide all the documentation for validation, including residue test methods.
Lieferumfang: Supplied with its own Certificate of Analysis. A detailed technical file (including: product specification, manufacturing process, shelf life, microbiological and analytical data) is available on request.
Verpackung: Available in 20, 200 and 1000 litre pack sizes.